| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013419.hg.1 |
ENST00000261383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013419.hg.1 |
ENST00000415178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013419.hg.1 |
NM_017539.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013419.hg.1 |
TC16000932.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600174215.hg.1 |
ENST00000261383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600174215.hg.1 |
ENST00000572640.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600174215.hg.1 |
NM_017539.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600174215.hg.1 |
ENST00000261383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600174215.hg.1 |
ENST00000572640.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600174215.hg.1 |
NM_017539.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600174215.hg.1 |
TC1600009671.hg.1 |
Clariom D TC |