Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185107.1 | NP_001172036.1 | 2 - 3 | 652 | 121 | |
| NM_001185108.1 | NP_001172037.1 | 2 - 3 | 652 | 121 | ||
| NM_001185115.1 | NP_001172044.1 | 2 - 3 | 652 | 121 | ||
| GenBank mRNA | AJ289115.1 | - | 2 - 3 | 359 | 121 | |
| AJ289116.1 | - | 2 - 3 | 359 | 121 | ||
| AJ289117.1 | - | 2 - 3 | 359 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001340.hg.1
Set Membership: |
|

CD1E
Genomic Map