Assay Details
Gene Symbol: KIRREL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005245305.4 | XP_005245362.1 | 8 - 9 | 1104 | 89 | |
GenBank mRNA | AK022708.1 | - | 4 - 5 | 449 | 89 | |
BC025268.1 | - | 3 - 4 | 295 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000359209.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000360089.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368172.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | ENST00000368173.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_001286349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010218.hg.1 | NM_018240.6 | Refseq |
