Assay Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031936.4 | NP_114142.3 | 1 - 2 | 81 | 117 | |
| XM_017002438.1 | XP_016857927.1 | 1 - 2 | 2414 | 117 | ||
| GenBank mRNA | AK290027.1 | - | 1 - 2 | 81 | 117 | |
| DA176879.1 | - | 1 - 2 | 81 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000946.hg
Additional Information:
This assay is in the 5'UTR region of the AK290027.1,NM_031936.4,XM_017002438.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014962.hg.1 | TC01000946.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014962.hg.1 | ENST00000286603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014962.hg.1 | ENST00000527748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014962.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014963.hg.1 | TC01000946.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014963.hg.1 | ENST00000286603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014963.hg.1 | ENST00000527748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014963.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014964.hg.1 | TC01000946.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014964.hg.1 | ENST00000286603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014964.hg.1 | ENST00000527748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014964.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166817.hg.1 | TC0100009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166819.hg.1 | TC0100009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166820.hg.1 | TC0100009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166815.hg.1 | TC0100009355.hg.1 | Clariom D TC |

GPR61
Genomic Map