Assay Details
| Gene Symbol: GSTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000849.4 | NP_000840.2 | 3 - 4 | 439 | 95 | |
| GenBank mRNA | AB209816.1 | - | 2 - 3 | 148 | 95 | |
| AK313444.1 | - | 2 - 3 | 157 | 95 | ||
| BC000088.2 | - | 2 - 3 | 144 | 3509784 | 95 | |
| BC008790.2 | - | 3 - 4 | 439 | 3630923 | 95 | |
| BE392471.1 | - | 3 - 4 | 420 | 96 | ||
| BT019945.1 | - | 2 - 3 | 129 | 95 | ||
| CR542108.1 | - | 2 - 3 | 129 | 95 | ||
| EF363096.1 | - | 2 - 3 | 146 | 95 | ||
| J05459.1 | - | 2 - 3 | 146 | 95 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
establishment of blood-nerve barrier
nitrobenzene metabolic process
xenobiotic catabolic process
response to estrogen
cellular detoxification of nitrogen compound
glutathione derivative biosynthetic process
glutathione transferase activity
enzyme binding
identical protein binding
protein homodimerization activity
glutathione binding
glutathione metabolic process
establishment of blood-nerve barrier
nitrobenzene metabolic process
xenobiotic catabolic process
response to estrogen
cellular detoxification of nitrogen compound
glutathione derivative biosynthetic process
glutathione transferase activity
enzyme binding
identical protein binding
protein homodimerization activity
glutathione binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014716.hg.1 | ENST00000412746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014716.hg.1 | ENST00000412746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000256594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000361066.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000476321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000486823.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000488824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | ENST00000540225.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NM_000849.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015211.hg.1 | NR_024537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014716.hg.1 | ENST00000412746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014716.hg.1 | ENST00000412746.1 | Ensembl |

GSTM3
Genomic Map