Assay Details
Gene Symbol: BICC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080512.2 | NP_001073981.1 | 2 - 3 | 239 | 76 | |
XM_011540185.2 | XP_011538487.1 | 3 - 4 | 485 | 76 | ||
XM_011540188.2 | XP_011538490.2 | 2 - 3 | 370 | 76 | ||
XM_017016677.1 | XP_016872166.1 | 3 - 4 | 6873 | 76 | ||
XM_017016678.1 | XP_016872167.1 | 4 - 5 | 390 | 76 | ||
GenBank mRNA | AY726586.1 | - | 2 - 3 | 184 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000362.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10004184.hg.1 | ENST00000373886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10004184.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10004184.hg.1 | TC10000362.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000152358.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000152358.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000152358.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000152358.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000152358.hg.1 | TC1000007703.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
