Assay Details
Gene Symbol: BICC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080512.2 | NP_001073981.1 | 4 - 5 | 383 | 104 | |
XM_005270169.4 | XP_005270226.1 | 3 - 4 | 3381 | 104 | ||
XM_011540185.2 | XP_011538487.1 | 5 - 6 | 629 | 104 | ||
XM_011540188.2 | XP_011538490.2 | 4 - 5 | 514 | 104 | ||
XM_011540189.2 | XP_011538491.1 | 3 - 4 | 280 | 104 | ||
XM_011540190.2 | XP_011538492.1 | 3 - 4 | 237 | 104 | ||
XM_017016677.1 | XP_016872166.1 | 5 - 6 | 7017 | 104 | ||
XM_017016678.1 | XP_016872167.1 | 6 - 7 | 534 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000362.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10004186.hg.1 | ENST00000373886.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10004186.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10004186.hg.1 | TC10000362.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000152360.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000152360.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000152360.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000152360.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000152360.hg.1 | TC1000007703.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000263103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000373886.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | ENST00000476684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000007703.hg.1 | NM_001080512.2 | Refseq |
