Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000476684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
ENST00000476684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004192.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004192.hg.1 |
ENST00000373886.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004192.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004192.hg.1 |
TC10000362.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000152369.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152369.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152369.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152369.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152369.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152369.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152369.hg.1 |
TC1000007703.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000263103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000373886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000476684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
ENST00000476684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007703.hg.1 |
NM_001080512.2 |
Refseq |