Assay Details
Gene Symbol: GUF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021927.2 | NP_068746.2 | 14 - 15 | 1924 | 123 | |
XM_005248122.3 | XP_005248179.1 | 13 - 14 | 1778 | 123 | ||
GenBank mRNA | AK023282.1 | - | 14 - 15 | 1874 | 123 | |
AK025248.1 | - | 6 - 7 | 704 | 123 | ||
BC036768.1 | - | 14 - 15 | 1885 | 5744283 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000268.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003893.hg.1 | TC04000268.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04003893.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003893.hg.1 | ENST00000506793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003893.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150539.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150539.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150539.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150539.hg.1 | ENST00000281543.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150539.hg.1 | ENST00000506793.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400150539.hg.1 | NM_021927.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400150539.hg.1 | TC0400007399.hg.1 | Clariom D TC |
