Assay Details
| Gene Symbol: GUF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021927.2 | NP_068746.2 | 15 - 16 | 2041 | 157 | |
| XM_005248122.3 | XP_005248179.1 | 14 - 15 | 1895 | 157 | ||
| GenBank mRNA | AK025248.1 | - | 7 - 8 | 821 | 157 | |
| AK225314.1 | - | 15 - 16 | 1996 | 157 | ||
| BC036768.1 | - | 15 - 16 | 2002 | 5744283 | 157 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000268.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04003894.hg.1 | TC04000268.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003894.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003894.hg.1 | ENST00000506793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003894.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150540.hg.1 | TC0400007399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |

GUF1
Genomic Map