Assay Details
| Gene Symbol: GUF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021927.2 | NP_068746.2 | 8 - 9 | 1152 | 73 | |
| XM_005248122.3 | XP_005248179.1 | 7 - 8 | 1006 | 73 | ||
| XM_011513732.2 | XP_011512034.1 | 8 - 9 | 1173 | 73 | ||
| GenBank mRNA | AK023282.1 | - | 8 - 9 | 1102 | 73 | |
| AK225314.1 | - | 8 - 9 | 1107 | 73 | ||
| AK308667.1 | - | 7 - 8 | 962 | 73 | ||
| BC036768.1 | - | 8 - 9 | 1113 | 5744283 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000268.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005248122.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003886.hg.1 | TC04000268.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003886.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003886.hg.1 | ENST00000506793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003886.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150532.hg.1 | TC0400007399.hg.1 | Clariom D TC |

GUF1
Genomic Map