Assay Details
| Gene Symbol: GUF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021927.2 | NP_068746.2 | 9 - 10 | 1281 | 144 | |
| XM_005248122.3 | XP_005248179.1 | 8 - 9 | 1135 | 144 | ||
| XM_011513732.2 | XP_011512034.1 | 9 - 10 | 1302 | 144 | ||
| GenBank mRNA | AK023282.1 | - | 9 - 10 | 1231 | 144 | |
| AK025248.1 | - | 1 - 2 | 61 | 144 | ||
| AK308667.1 | - | 8 - 9 | 1091 | 144 | ||
| BC036768.1 | - | 9 - 10 | 1242 | 5744283 | 144 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000268.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK025248.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04003887.hg.1 | TC04000268.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003887.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003887.hg.1 | ENST00000506793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003887.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150533.hg.1 | TC0400007399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000281543.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000506793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000511493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | ENST00000513775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007399.hg.1 | NM_021927.2 | Refseq |

GUF1
Genomic Map