Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009265.hg.1 |
TC06000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009265.hg.1 |
ENST00000369687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009265.hg.1 |
ENST00000369689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009265.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160197.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160197.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160197.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160197.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160197.hg.1 |
TC0600008674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008674.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160195.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160195.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160195.hg.1 |
TC0600008674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
ENST00000369687.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
ENST00000369689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
NM_001009994.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
NR_103525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160200.hg.1 |
TC0600008674.hg.1 |
Clariom D TC |