Assay Details
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168398.1 | NP_001161870.1 | 3 - 4 | 431 | 78 | |
NM_006416.4 | NP_006407.1 | 3 - 4 | 431 | 78 | ||
GenBank mRNA | AJ810302.1 | - | 3 - 4 | 365 | 78 | |
AJ810303.1 | - | 3 - 4 | 367 | 78 | ||
AJ851888.1 | - | 3 - 4 | 352 | 78 | ||
AJ851889.1 | - | 3 - 4 | 352 | 78 | ||
AK299432.1 | - | 3 - 4 | 379 | 78 | ||
AL122119.1 | - | 3 - 4 | 427 | 78 | ||
BC017807.1 | - | 3 - 4 | 379 | 4690513 | 78 | |
BG699364.1 | - | 4 - 5 | 480 | 78 | ||
D87969.1 | - | 3 - 4 | 379 | 78 | ||
DA210713.1 | - | 3 - 4 | 437 | 78 | ||
DB095595.1 | - | 3 - 4 | 431 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06004096.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | TC06004096.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | ENST00000369544.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | ENST00000369552.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | ENST00000369556.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | ENST00000369557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | ENST00000429605.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | ENST00000464978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | ENST00000544441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06009455.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199163.hg.1 | TC0600014154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
