Assay Details
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168398.1 | NP_001161870.1 | 5 - 6 | 653 | 69 | |
GenBank mRNA | AJ851888.1 | - | 5 - 6 | 574 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06004096.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | TC06004096.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | ENST00000369544.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | ENST00000369552.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | ENST00000369556.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | ENST00000369557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | ENST00000464978.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | ENST00000544441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06009466.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600199167.hg.1 | TC0600014154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369552.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369556.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000369557.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000464978.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000544441.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | ENST00000622775.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_001168398.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014154.hg.1 | NM_006416.4 | Refseq |
