Assay Details
| Gene Symbol: PODN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153703.4 | NP_714914.2 | 5 - 6 | 895 | 59 | |
| XM_005270457.3 | XP_005270514.1 | 6 - 7 | 924 | 59 | ||
| XM_017000294.1 | XP_016855783.1 | 5 - 6 | 1245 | 59 | ||
| GenBank mRNA | AK296649.1 | - | 6 - 7 | 790 | 59 | |
| AK299375.1 | - | 7 - 8 | 980 | 59 | ||
| AK314096.1 | - | 5 - 6 | 489 | 59 | ||
| AY313607.1 | - | 5 - 6 | 734 | 59 | ||
| AY358294.1 | - | 7 - 8 | 1011 | 59 | ||
| BC028292.1 | - | 5 - 6 | 718 | 4938380 | 59 | |
| BC030608.2 | - | 5 - 6 | 736 | 4826932 | 59 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000649.hg
Additional Information:
This assay is in the 5'UTR region of the AK314096.1,BC028292.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000312553.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000371500.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000395871.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000471285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000490650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000490650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000490650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000490650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000490650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | ENST00000618387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199080.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199081.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_001199082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008341.hg.1 | NM_153703.4 | Refseq |

PODN
Genomic Map