Assay Details
| Gene Symbol: SP3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036697.1 | - | 1 | 1563 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474653 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474694 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474628 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16656618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474732 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474667 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16656593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474673 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474744 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16656616 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474611 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474655 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474670 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474729 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474666 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474616 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16656596 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026344 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16656599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026295 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474683 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026339 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474657 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474662 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474719 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474638 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474614 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474733 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474663 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474641 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474625 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474636 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474619 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474626 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474637 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16656617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026341 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09026321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474612 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29474678 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 411127 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | AK026669.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | BC021048.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | BC029412.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | BC105588.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | CR604424.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | D50663.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | NM_006519.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 100629 | U56255.1 | genbank_mrna_human |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3833478 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P203668 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 44581 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 39120 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3833478 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P203668 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P203668 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33438866 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33439038 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P930125 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P33439038 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7972249 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7972248 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7972250 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7972247 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 216403_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216403_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216403_at | - | - |
| Human Genome U133A 2.0 Array | 216403_at | - | - |
| Human Genome U133A 2.0 Array | 216403_PM_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw531694 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216403_at7 | - | - |
| Human Genome U133 Set | 216403_at | - | - |
| Human Genome U133 Set | 216403_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 110698 | - | - |

SP3P
Genomic Map