Assay Details
| Gene Symbol: SLC12A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097808.1 | - | 5 - 6 | 590 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK097808.1 transcripts.
Set Membership: |
|

SLC12A4
Genomic Map