| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001649.hg.1 |
ENST00000379487.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001649.hg.1 |
ENST00000542082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001649.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001649.hg.1 |
TC13000141.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1300147235.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147235.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147235.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147235.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147235.hg.1 |
TC1300006965.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |