Assay Details
| Gene Symbol: NFYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002505.4 | NP_002496.1 | 3 - 4 | 347 | 70 | |
| GenBank mRNA | GQ167198.1 | - | 2 - 3 | 166 | 70 | |
| X59711.1 | - | 3 - 4 | 343 | 70 | ||
More Information
Set Membership: |
|

NFYA
Genomic Map