Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052733.hg.1 |
TC01003371.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052733.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052733.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222039.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222039.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222039.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222039.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222039.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222039.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222039.hg.1 |
TC0100016065.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000368168.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
ENST00000451207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016065.hg.1 |
NM_001764.2 |
Refseq |