Assay Details
Gene Symbol: PDCD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014976.1 | NP_055791.1 | 27 - 28 | 4116 | 73 | |
XM_005269647.3 | XP_005269704.1 | 27 - 28 | 4156 | 73 | ||
XM_011539538.2 | XP_011537840.1 | 27 - 28 | 4104 | 73 | ||
XM_011539539.2 | XP_011537841.1 | 27 - 28 | 4104 | 73 | ||
XM_011539540.1 | XP_011537842.1 | 26 - 27 | 3970 | 73 | ||
GenBank mRNA | BC049838.1 | - | 27 - 28 | 4117 | 73 | |
BC064486.1 | - | 27 - 28 | 4109 | 73 | ||
BC080560.1 | - | 27 - 28 | 4109 | 73 | ||
BC111040.1 | - | 27 - 28 | 4071 | 73 | ||
D80007.1 | - | 27 - 28 | 4069 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000773.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10009232.hg.1 | ENST00000369797.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009232.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009232.hg.1 | TC10000773.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000490787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000490787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160078.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160078.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160078.hg.1 | ENST00000369797.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000160078.hg.1 | NM_014976.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000160078.hg.1 | TC1000008770.hg.1 | Clariom D TC |
