Assay Details
| Gene Symbol: PSMG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020232.4 | NP_064617.2 | 4 - 5 | 527 | 149 | |
| NM_147163.1 | NP_671692.1 | 4 - 5 | 385 | 149 | ||
| GenBank mRNA | AF068295.1 | - | 4 - 5 | 385 | 149 | |
| AF161378.1 | - | 4 - 5 | 385 | 149 | ||
| AF276707.1 | - | 4 - 5 | 1087 | 149 | ||
| AK057005.1 | - | 4 - 5 | 459 | 149 | ||
| AK225882.1 | - | 4 - 5 | 494 | 149 | ||
| BC013356.2 | - | 4 - 5 | 450 | 3640209 | 149 | |
| CR457181.1 | - | 3 - 4 | 287 | 149 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18001006.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000719.hg.1 | TC18001006.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000719.hg.1 | ENST00000317615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000719.hg.1 | ENST00000539202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000719.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000719.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000400512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000400512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585853.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585853.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000585853.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000589405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000589405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000589405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | TC1800006739.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146144.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000317615.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000400512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000400512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000400514.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585331.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585853.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585853.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000585853.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000586587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000588824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000589405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000589405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000589405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | ENST00000590217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_020232.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006739.hg.1 | NM_147163.1 | Refseq |

PSMG2
Genomic Map