Assay Details
| Gene Symbol: TM9SF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020123.3 | NP_064508.3 | 1 - 2 | 316 | 140 | |
| GenBank mRNA | AF160213.1 | - | 1 - 2 | 115 | 137 | |
| AF269150.1 | - | 1 - 2 | 316 | 140 | ||
| AK314348.1 | - | 1 - 2 | 282 | 140 | ||
| BC136788.1 | - | 1 - 2 | 316 | 9020778 | 140 | |
| BC136789.1 | - | 1 - 2 | 316 | 9020779 | 140 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001558.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10019784.hg.1 | ENST00000371142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019784.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019784.hg.1 | TC10001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019786.hg.1 | ENST00000371142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019786.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019786.hg.1 | TC10001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019787.hg.1 | ENST00000371142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019787.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019787.hg.1 | TC10001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178344.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178344.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178344.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178344.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178344.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178344.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178344.hg.1 | TC1000011522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000178345.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178345.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178345.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178345.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178345.hg.1 | TC1000011522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178342.hg.1 | TC1000011522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000178346.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178346.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178346.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178346.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178346.hg.1 | TC1000011522.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 166068 | - | - |

TM9SF3
Genomic Map