Assay Details
| Gene Symbol: TM9SF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020123.3 | NP_064508.3 | 5 - 6 | 881 | 94 | |
| XM_011539976.2 | XP_011538278.1 | 5 - 6 | 961 | 94 | ||
| XM_011539977.1 | XP_011538279.1 | 5 - 6 | 961 | 94 | ||
| GenBank mRNA | AF116347.1 | - | 5 - 6 | 631 | 94 | |
| AF269150.1 | - | 5 - 6 | 881 | 94 | ||
| AK027788.1 | - | 4 - 5 | 537 | 94 | ||
| AK074824.1 | - | 2 - 3 | 172 | 94 | ||
| AK314348.1 | - | 5 - 6 | 847 | 94 | ||
| AY358819.1 | - | 4 - 5 | 551 | 94 | ||
| BC020959.1 | - | 4 - 5 | 461 | 94 | ||
| BC136788.1 | - | 5 - 6 | 881 | 9020778 | 94 | |
| BC136789.1 | - | 5 - 6 | 881 | 9020779 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001558.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019778.hg.1 | ENST00000371142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019778.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019778.hg.1 | TC10001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019779.hg.1 | ENST00000371142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019779.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019779.hg.1 | TC10001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019780.hg.1 | ENST00000371142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019780.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019780.hg.1 | TC10001558.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178338.hg.1 | TC1000011522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000475401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000485093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | ENST00000490192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011522.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178336.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178336.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178336.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178336.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178336.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178336.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178336.hg.1 | TC1000011522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | ENST00000371142.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | ENST00000443638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | ENST00000464654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | NM_020123.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000178337.hg.1 | TC1000011522.hg.1 | Clariom D TC |

TM9SF3
Genomic Map