Assay Details
Gene Symbol: MEPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291183.1 | NP_001278112.1 | 3 - 4 | 207 | 131 | |
XM_006714278.2 | XP_006714341.1 | 4 - 5 | 290 | 131 | ||
GenBank mRNA | DQ854717.1 | - | 3 - 4 | 170 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000361056.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000395102.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000424957.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000497649.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000508016.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000511670.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000515821.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000540395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000560249.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184694.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001291183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_020203.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000361056.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000395102.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000424957.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000497649.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000508016.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000511670.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000515821.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000540395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | ENST00000560249.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184694.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001184697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_001291183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | NM_020203.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155583.hg.1 | TC0400008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000361056.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000424957.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000540395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000560249.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184694.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001291183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_020203.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000361056.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000424957.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000540395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | ENST00000560249.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184694.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001184697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_001291183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | NM_020203.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155589.hg.1 | TC0400008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000361056.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000395102.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000424957.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000497649.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000508016.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000540395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000560249.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184694.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001291183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_020203.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000361056.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000395102.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000424957.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000497649.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000508016.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000540395.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | ENST00000560249.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184694.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184695.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184696.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001184697.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_001291183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | NM_020203.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400155588.hg.1 | TC0400008088.hg.1 | Clariom D TC |
