Assay Details
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001711.5 | NP_001702.1 | 2 - 3 | 477 | 77 | |
XM_017029724.1 | XP_016885213.1 | 1 - 2 | 256 | 77 | ||
GenBank mRNA | AK092954.1 | - | 2 - 3 | 385 | 77 | |
AK094059.1 | - | 3 - 4 | 286 | 77 | ||
AK222484.1 | - | 2 - 3 | 386 | 77 | ||
AK223302.1 | - | 2 - 3 | 357 | 77 | ||
AK293268.1 | - | 2 - 3 | 413 | 77 | ||
AK293289.1 | - | 2 - 3 | 312 | 77 | ||
AK298756.1 | - | 1 - 2 | 186 | 77 | ||
AU142113.1 | - | 2 - 3 | 413 | 77 | ||
BC002416.2 | - | 2 - 3 | 370 | 3162633 | 77 | |
BC004244.1 | - | 2 - 3 | 380 | 3503374 | 77 | |
BT007323.1 | - | 1 - 2 | 241 | 77 | ||
DC299956.1 | - | 2 - 3 | 477 | 77 | ||
DC419932.1 | - | 2 - 3 | 457 | 77 | ||
DC425011.1 | - | 2 - 3 | 438 | 77 | ||
J04599.1 | - | 2 - 3 | 361 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000750.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
negative regulation of protein kinase activity
biological_process
cytokine-mediated signaling pathway
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
extracellular matrix structural constituent
glycosaminoglycan binding
extracellular matrix binding
blood vessel remodeling
negative regulation of protein kinase activity
biological_process
cytokine-mediated signaling pathway
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
extracellular matrix structural constituent
glycosaminoglycan binding
extracellular matrix binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X009747.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009747.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009747.hg.1 | ENST00000370204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009747.hg.1 | ENST00000430380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009747.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009747.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009745.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009745.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009745.hg.1 | ENST00000370204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009745.hg.1 | ENST00000430380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009745.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009744.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009744.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009744.hg.1 | ENST00000370204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009744.hg.1 | ENST00000430380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009744.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009744.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159777.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159776.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159774.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
