Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002665.hg.1 |
ENST00000221980.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002665.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002665.hg.1 |
TC19000176.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000588912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000588912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
ENST00000588912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900149644.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149644.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149644.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900149644.hg.1 |
TC1900006979.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900149644.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149644.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900149644.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000221980.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000586480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000587398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000588912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000588912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
ENST00000588912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006979.hg.1 |
NM_003259.3 |
Refseq |