Assay Details
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001711.5 | NP_001702.1 | 3 - 4 | 590 | 89 | |
XM_017029724.1 | XP_016885213.1 | 2 - 3 | 369 | 89 | ||
GenBank mRNA | AK093023.1 | - | 3 - 4 | 424 | 89 | |
AK094059.1 | - | 4 - 5 | 399 | 89 | ||
AK222484.1 | - | 3 - 4 | 499 | 89 | ||
AK223302.1 | - | 3 - 4 | 470 | 89 | ||
AK291955.1 | - | 3 - 4 | 501 | 89 | ||
AK293289.1 | - | 3 - 4 | 425 | 89 | ||
AU142113.1 | - | 3 - 4 | 526 | 89 | ||
BC002416.2 | - | 3 - 4 | 483 | 3162633 | 89 | |
BC004244.1 | - | 3 - 4 | 493 | 3503374 | 89 | |
BT007323.1 | - | 2 - 3 | 354 | 89 | ||
J04599.1 | - | 3 - 4 | 474 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000750.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
negative regulation of protein kinase activity
biological_process
cytokine-mediated signaling pathway
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
extracellular matrix structural constituent
glycosaminoglycan binding
extracellular matrix binding
blood vessel remodeling
negative regulation of protein kinase activity
biological_process
cytokine-mediated signaling pathway
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
extracellular matrix structural constituent
glycosaminoglycan binding
extracellular matrix binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X009751.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009751.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009751.hg.1 | ENST00000370204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009751.hg.1 | ENST00000430380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009751.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009751.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009752.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009752.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009752.hg.1 | ENST00000370204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009752.hg.1 | ENST00000430380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009752.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009752.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009748.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009748.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009748.hg.1 | ENST00000370204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009748.hg.1 | ENST00000430380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009748.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009748.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159781.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159780.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
