Assay Details
| Gene Symbol: PDCD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014976.1 | NP_055791.1 | 31 - 32 | 4789 | 67 | |
| XM_011539539.2 | XP_011537841.1 | 31 - 32 | 4777 | 67 | ||
| GenBank mRNA | D80007.1 | - | 31 - 32 | 4742 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000773.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000490787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009239.hg.1 | ENST00000369797.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009239.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009239.hg.1 | TC10000773.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000466959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000471061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000478543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000490787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | ENST00000493610.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008770.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160084.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160084.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160084.hg.1 | ENST00000369797.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160084.hg.1 | NM_014976.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160084.hg.1 | TC1000008770.hg.1 | Clariom D TC |

PDCD11
Genomic Map