Assay Details
| Gene Symbol: AS3MT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020682.3 | NP_065733.2 | 2 - 3 | 176 | 73 | |
| GenBank mRNA | AF226730.1 | - | 2 - 3 | 113 | 73 | |
| AK057833.1 | - | 2 - 3 | 195 | 73 | ||
| AK225972.1 | - | 2 - 3 | 195 | 73 | ||
| AK309737.1 | - | 2 - 3 | 108 | 73 | ||
| BC119637.1 | - | 2 - 3 | 80 | 73 | ||
| BC119638.2 | - | 2 - 3 | 80 | 73 | ||
| DA368970.1 | - | 2 - 3 | 176 | 73 | ||
| Gene Symbol: BORCS7-ASMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037644.1 | 6 - 7 | 441 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002948.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009128.hg.1 | TC10002948.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009128.hg.1 | ENST00000369880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009128.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186248.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186248.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186248.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186248.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186248.hg.1 | TC1000012497.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186221.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186221.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186221.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186221.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186221.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186221.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186221.hg.1 | TC1000012496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186249.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186249.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186249.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186249.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186249.hg.1 | TC1000012497.hg.1 | Clariom D TC |

AS3MT
Genomic Map