Assay Details
| Gene Symbol: HPGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145816.2 | NP_001139288.1 | 5 - 6 | 944 | 70 | |
| GenBank mRNA | AK300940.1 | - | 5 - 6 | 533 | 70 | |
| X82460.1 | - | 5 - 6 | 497 | 70 | ||
More Information
Set Membership: |
|

HPGD
Genomic Map