Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000480404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000480404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011464.hg.1 |
TC04000829.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011464.hg.1 |
ENST00000261510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011464.hg.1 |
ENST00000402744.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011464.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163398.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163398.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163398.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163398.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163398.hg.1 |
TC0400009223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000402744.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000431967.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000480404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000480404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000511992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
ENST00000513982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009223.hg.1 |
NM_001873.3 |
Refseq |