Assay Details
| Gene Symbol: UGT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128174.2 | NP_001121646.1 | 1 - 2 | 578 | 106 | |
| GenBank mRNA | AK127970.1 | - | 1 - 2 | 508 | 106 | |
| AK225763.1 | - | 1 - 2 | 171 | 106 | ||
| BC075069.2 | - | 1 - 2 | 50 | 30915590 | 106 | |
| DA295156.1 | - | 1 - 2 | 517 | 106 | ||
| U62899.1 | - | 1 - 2 | 229 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000590.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK127970.1,BC075069.2,NM_001128174.2,U62899.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
protein localization to paranode region of axon
galactosylceramide biosynthetic process
glycosphingolipid metabolic process
cytoskeleton organization
central nervous system development
peripheral nervous system development
flavonoid biosynthetic process
paranodal junction assembly
neuron projection morphogenesis
flavonoid glucuronidation
2-hydroxyacylsphingosine 1-beta-galactosyltransferase activity
UDP-galactose:glucosylceramide beta-1,4-galactosyltransferase activity
glucuronosyltransferase activity
N-acylsphingosine galactosyltransferase activity
protein localization to paranode region of axon
galactosylceramide biosynthetic process
glycosphingolipid metabolic process
cytoskeleton organization
central nervous system development
peripheral nervous system development
flavonoid biosynthetic process
paranodal junction assembly
neuron projection morphogenesis
flavonoid glucuronidation
2-hydroxyacylsphingosine 1-beta-galactosyltransferase activity
UDP-galactose:glucosylceramide beta-1,4-galactosyltransferase activity
glucuronosyltransferase activity
N-acylsphingosine galactosyltransferase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04008642.hg.1 | TC04000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008642.hg.1 | ENST00000310836.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008642.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008643.hg.1 | TC04000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008643.hg.1 | ENST00000310836.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008643.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008647.hg.1 | TC04000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008647.hg.1 | ENST00000310836.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008647.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008647.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008647.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008641.hg.1 | TC04000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008641.hg.1 | ENST00000310836.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008641.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008646.hg.1 | TC04000590.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008646.hg.1 | ENST00000310836.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008646.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008646.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008646.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158244.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158244.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158244.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158244.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158244.hg.1 | TC0400008483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400158245.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158245.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158245.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158245.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158245.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158245.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158245.hg.1 | TC0400008483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008483.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158253.hg.1 | TC0400008483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | ENST00000394511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | NM_003360.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158252.hg.1 | TC0400008483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400158246.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158246.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158246.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158246.hg.1 | ENST00000310836.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158246.hg.1 | ENST00000507710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158246.hg.1 | NM_001128174.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158246.hg.1 | TC0400008483.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 180976 | - | - |

UGT8
Genomic Map