Assay Details
| Gene Symbol: GRAMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005261396.3 | XP_005261453.1 | 1 - 2 | 98 | 64 | |
| XM_011530013.2 | XP_011528315.2 | 1 - 2 | 107 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005261396.3, XM_011530013.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000490378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | ENST00000490378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000361034.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000406902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000408031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000431155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000447351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000456069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000490378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | ENST00000490378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007672.hg.1 | NM_015124.4 | Refseq |

GRAMD4
Genomic Map