Assay Details
Gene Symbol: RDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172037.4 | NP_742034.1 | 2 - 3 | 1210 | 86 | |
GenBank mRNA | AF456765.1 | - | 2 - 3 | 782 | 86 | |
AY358270.1 | - | 2 - 3 | 602 | 86 | ||
BC067131.1 | - | 2 - 3 | 710 | 6153778 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000482.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
metanephros development
in utero embryonic development
retinoic acid biosynthetic process
visual perception
gonad development
neural crest cell development
embryonic camera-type eye development
embryonic forelimb morphogenesis
retinol metabolic process
retinal metabolic process
ear development
nose development
embryonic viscerocranium morphogenesis
oxidation-reduction process
primary lung bud formation
bud elongation involved in lung branching
retinol dehydrogenase activity
NADP-retinol dehydrogenase activity
retinoid metabolic process
metanephros development
in utero embryonic development
retinoic acid biosynthetic process
visual perception
gonad development
neural crest cell development
embryonic camera-type eye development
embryonic forelimb morphogenesis
retinol metabolic process
retinal metabolic process
ear development
nose development
embryonic viscerocranium morphogenesis
oxidation-reduction process
primary lung bud formation
bud elongation involved in lung branching
retinol dehydrogenase activity
NADP-retinol dehydrogenase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08006662.hg.1 | TC08000482.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08006662.hg.1 | ENST00000240285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08006662.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08006662.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800154855.hg.1 | TC0800007957.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000240285.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000518870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000519380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | ENST00000521928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007957.hg.1 | NM_172037.4 | Refseq |
