Assay Details
| Gene Symbol: JRKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261833.1 | NP_001248762.1 | 2 | 98 | ||
| NM_003772.3 | NP_003763.2 | 1 | 98 | |||
| GenBank mRNA | AF004715.1 | - | 1 | 98 | ||
| CA503357.1 | - | 1 | 468 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000936.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11011694.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011694.hg.1 | ENST00000458427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011694.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011694.hg.1 | TC11000936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011697.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011697.hg.1 | ENST00000458427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011697.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011697.hg.1 | TC11000936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011693.hg.1 | ENST00000458427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011693.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011693.hg.1 | TC11000936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011695.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011695.hg.1 | ENST00000458427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011695.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011695.hg.1 | TC11000936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011692.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011692.hg.1 | ENST00000458427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011692.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011692.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011692.hg.1 | TC11000936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011696.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011696.hg.1 | ENST00000458427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011696.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11011696.hg.1 | TC11000936.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100165361.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165361.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165361.hg.1 | TC1100008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100165363.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165363.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165363.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165363.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165363.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165363.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165363.hg.1 | TC1100008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100165364.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165364.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165364.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165364.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165364.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165364.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165364.hg.1 | TC1100008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100165365.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165365.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165365.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165365.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165365.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165365.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165365.hg.1 | TC1100008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100165362.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165362.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165362.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165362.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165362.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165362.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165362.hg.1 | TC1100008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100165360.hg.1 | TC1100008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000332349.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | ENST00000546177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_001261833.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_003772.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008856.hg.1 | NM_003772.3 | Refseq |

JRKL
Genomic Map