Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021699.hg.1 |
TC03001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021699.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021699.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021698.hg.1 |
TC03001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021698.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021698.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021700.hg.1 |
TC03001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021700.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021700.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177452.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177452.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177452.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177452.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177452.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177452.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177452.hg.1 |
TC0300010643.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300177451.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177451.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177451.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177451.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177451.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177451.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177451.hg.1 |
TC0300010643.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300177450.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177450.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177450.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177450.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177450.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177450.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177450.hg.1 |
TC0300010643.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300177453.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177453.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177453.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177453.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177453.hg.1 |
TC0300010643.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300177454.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177454.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177454.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177454.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177454.hg.1 |
TC0300010643.hg.1 |
Clariom D TC |