Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021696.hg.1 |
TC03001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021696.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021696.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010643.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
ENST00000205636.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
ENST00000478886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
ENST00000495177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
NM_017801.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300177448.hg.1 |
TC0300010643.hg.1 |
Clariom D TC |