Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008426.hg.1 |
ENST00000155674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008426.hg.1 |
ENST00000336737.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008426.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008426.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008426.hg.1 |
TC22000471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000463033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000463033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
TC2200007892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156071.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000155674.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000336737.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000399852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000463033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000463033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000477157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000480451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
ENST00000486462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_017829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007892.hg.1 |
NM_033070.2 |
Refseq |