Assay Details
| Gene Symbol: CECR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017829.5 | NP_060299.4 | 5 - 6 | 530 | 104 | |
| NM_033070.2 | NP_149061.1 | 5 - 6 | 605 | 104 | ||
| XM_005261236.3 | XP_005261293.1 | 5 - 6 | 583 | 104 | ||
| XM_011546126.2 | XP_011544428.1 | 4 - 5 | 401 | 104 | ||
| XM_011546127.1 | XP_011544429.1 | 5 - 6 | 1020 | 104 | ||
| XM_017028781.1 | XP_016884270.1 | 5 - 6 | 1072 | 104 | ||
| GenBank mRNA | AF273270.1 | - | 5 - 6 | 530 | 104 | |
| AF273271.1 | - | 5 - 6 | 590 | 104 | ||
| AK000461.1 | - | 5 - 6 | 603 | 104 | ||
| AK001034.1 | - | 5 - 6 | 583 | 104 | ||
| AK315156.1 | - | 5 - 6 | 591 | 104 | ||
| BC042540.1 | - | 5 - 6 | 589 | 4342582 | 104 | |
| BM558767.1 | - | 4 - 5 | 409 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000471.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000463033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000463033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008424.hg.1 | ENST00000155674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008424.hg.1 | ENST00000336737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008424.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008424.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008424.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008424.hg.1 | TC22000471.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008420.hg.1 | ENST00000155674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008420.hg.1 | ENST00000336737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008420.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008420.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008420.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008420.hg.1 | TC22000471.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000463033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000463033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000480451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007892.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | TC2200007892.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156069.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | TC2200007892.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | NM_033070.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000155674.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000336737.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000399852.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000477157.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | ENST00000486462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | NM_017829.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200156065.hg.1 | NM_033070.2 | Refseq |

CECR5
Genomic Map