Assay Details
Gene Symbol: CDC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001255.2 | NP_001246.2 | 5 - 6 | 661 | 64 | |
GenBank mRNA | AF099644.1 | - | 4 - 5 | 604 | 64 | |
AK225504.1 | - | 5 - 6 | 640 | 64 | ||
AK302539.1 | - | 3 - 4 | 893 | 64 | ||
AK303844.1 | - | 6 - 7 | 616 | 64 | ||
AK312780.1 | - | 5 - 6 | 640 | 64 | ||
BC000624.2 | - | 4 - 5 | 668 | 3346682 | 64 | |
BC001088.2 | - | 5 - 6 | 661 | 2964686 | 64 | |
BC006272.2 | - | 4 - 5 | 603 | 3948819 | 64 | |
BC009425.1 | - | 5 - 6 | 643 | 2959460 | 64 | |
BC009426.1 | - | 5 - 6 | 643 | 2959596 | 64 | |
BC010044.2 | - | 5 - 6 | 652 | 3634656 | 64 | |
BC012803.2 | - | 5 - 6 | 643 | 2959588 | 64 | |
BC012827.2 | - | 5 - 6 | 661 | 2964686 | 64 | |
BC013303.2 | - | 5 - 6 | 643 | 2959588 | 64 | |
BC015998.1 | - | 5 - 6 | 639 | 4306426 | 64 | |
BC024257.1 | - | 5 - 6 | 643 | 2959460 | 64 | |
BC031294.1 | - | 5 - 6 | 643 | 2959487 | 64 | |
BC110321.1 | - | 4 - 5 | 624 | 5496957 | 64 | |
BT007388.1 | - | 4 - 5 | 560 | 64 | ||
U05340.1 | - | 5 - 6 | 670 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
