Assay Details
| Gene Symbol: TNFRSF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001066.2 | NP_001057.1 | 1 - 2 | 173 | 74 | |
| XM_011542060.2 | XP_011540362.1 | 1 - 2 | 228 | 74 | ||
| XM_011542063.2 | XP_011540365.1 | 1 - 2 | 228 | 74 | ||
| XM_017002211.1 | XP_016857700.1 | 1 - 2 | 228 | 74 | ||
| GenBank mRNA | AY148473.1 | - | 1 - 2 | 90 | 74 | |
| BC052977.1 | - | 1 - 2 | 171 | 6198614 | 74 | |
| BT019927.1 | - | 1 - 2 | 84 | 74 | ||
| EU826615.1 | - | 1 - 2 | 84 | 74 | ||
| M32315.1 | - | 1 - 2 | 173 | 74 | ||
| M55994.1 | - | 1 - 2 | 176 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000155.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
immune response
multicellular organism development
aging
intrinsic apoptotic signaling pathway in response to DNA damage
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
regulation of apoptotic process
positive regulation of MAPK cascade
negative regulation of inflammatory response
RNA destabilization
positive regulation of membrane protein ectodomain proteolysis
cellular response to lipopolysaccharide
cellular response to growth factor stimulus
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
tumor necrosis factor-activated receptor activity
protein binding
ubiquitin protein ligase binding
inflammatory response
immune response
multicellular organism development
aging
intrinsic apoptotic signaling pathway in response to DNA damage
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
regulation of apoptotic process
positive regulation of MAPK cascade
negative regulation of inflammatory response
RNA destabilization
positive regulation of membrane protein ectodomain proteolysis
cellular response to lipopolysaccharide
cellular response to growth factor stimulus
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
tumor necrosis factor-activated receptor activity
protein binding
ubiquitin protein ligase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01002583.hg.1 | TC01000155.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002583.hg.1 | ENST00000376259.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002583.hg.1 | ENST00000400863.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002583.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002583.hg.1 | ENST00000536782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002583.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002584.hg.1 | TC01000155.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002584.hg.1 | ENST00000376259.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002584.hg.1 | ENST00000400863.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002584.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002584.hg.1 | ENST00000536782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002584.hg.1 | ENST00000539840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002584.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000584158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NR_039775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002587.hg.1 | TC01000155.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002587.hg.1 | ENST00000376259.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002587.hg.1 | ENST00000400863.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002587.hg.1 | ENST00000536782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002587.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147925.hg.1 | TC0100006881.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000584158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NR_039775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147924.hg.1 | TC0100006881.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100147926.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147926.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147926.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147926.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147926.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147926.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147926.hg.1 | TC0100006881.hg.1 | Clariom D TC |

TNFRSF1B
Genomic Map