Assay Details
Gene Symbol: TNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022093.1 | NP_071376.1 | 13 - 14 | 3162 | 130 | |
XM_017002048.1 | XP_016857537.1 | 13 - 14 | 3167 | 130 | ||
XM_017002049.1 | XP_016857538.1 | 12 - 13 | 2904 | 130 | ||
GenBank mRNA | AK127044.1 | - | 13 - 14 | 3162 | 130 | |
AK127654.1 | - | 3 - 4 | 582 | 130 | ||
AL049689.1 | - | 12 - 13 | 3034 | 130 | ||
BC136619.1 | - | 12 - 13 | 3079 | 9020609 | 130 | |
BC144305.1 | - | 11 - 12 | 2548 | 9052825 | 130 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001527.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01023627.hg.1 | TC01001527.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01023627.hg.1 | ENST00000239462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01023627.hg.1 | ENST00000539081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01023627.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100178592.hg.1 | TC0100010689.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000239462.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000621086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | ENST00000622870.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010689.hg.1 | NM_022093.1 | Refseq |
