Assay Details
Gene Symbol: HIBCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198047.2 | NP_932164.1 | 12 - 13 | 1297 | 125 | |
GenBank mRNA | BC005190.1 | - | 12 - 13 | 1074 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
