Assay Details
Gene Symbol: DEPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022783.3 | NP_073620.2 | 1 - 2 | 310 | 103 | |
GenBank mRNA | AK022490.1 | - | 1 - 2 | 198 | 103 | |
AK023916.1 | - | 1 - 2 | 94 | 103 | ||
AK225168.1 | - | 1 - 2 | 243 | 103 | ||
AK225675.1 | - | 1 - 2 | 216 | 103 | ||
AK315175.1 | - | 1 - 2 | 248 | 103 | ||
AL136678.1 | - | 1 - 2 | 240 | 103 | ||
BC012040.1 | - | 1 - 2 | 154 | 3855457 | 103 | |
BC024746.1 | - | 1 - 2 | 210 | 5112749 | 103 | |
DA622978.1 | - | 1 - 2 | 309 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000698.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK023916.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08009420.hg.1 | TC08000698.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009420.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009420.hg.1 | ENST00000500705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009420.hg.1 | ENST00000523492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009420.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08009418.hg.1 | TC08000698.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009418.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009418.hg.1 | ENST00000500705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009418.hg.1 | ENST00000523492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009418.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08009417.hg.1 | TC08000698.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009417.hg.1 | ENST00000500705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009417.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08009419.hg.1 | TC08000698.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009419.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009419.hg.1 | ENST00000500705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009419.hg.1 | ENST00000523492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009419.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08009422.hg.1 | TC08000698.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009422.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009422.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159424.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159424.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159424.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159424.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159424.hg.1 | TC0800008674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159427.hg.1 | TC0800008674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159425.hg.1 | TC0800008674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159426.hg.1 | TC0800008674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159428.hg.1 | TC0800008674.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000518057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | ENST00000523492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_001283012.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008674.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159430.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159430.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159430.hg.1 | ENST00000286234.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800159430.hg.1 | NM_022783.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800159430.hg.1 | TC0800008674.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 105011 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 205521 | - | - |
