Assay Details
| Gene Symbol: DDX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004398.3 | NP_004389.2 | 15 - 16 | 2370 | 99 | |
| GenBank mRNA | AB040537.1 | - | 15 - 16 | 2306 | 99 | |
| AB209884.1 | - | 15 - 16 | 2273 | 99 | ||
| AK296752.1 | - | 13 - 14 | 2044 | 99 | ||
| AK315216.1 | - | 15 - 16 | 2301 | 99 | ||
| BC091521.1 | - | 15 - 16 | 2318 | 30388370 | 99 | |
| BC093654.1 | - | 15 - 16 | 2274 | 7939499 | 99 | |
| BC093656.1 | - | 15 - 16 | 2274 | 7939501 | 99 | |
| U28042.1 | - | 15 - 16 | 2306 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000981.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11012275.hg.1 | ENST00000322536.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012275.hg.1 | ENST00000456020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012275.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012275.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012275.hg.1 | TC11000981.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012276.hg.1 | ENST00000322536.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012276.hg.1 | ENST00000456020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012276.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012276.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012276.hg.1 | TC11000981.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000524979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000530116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000533198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | ENST00000534774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008991.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100166498.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100166498.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100166498.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100166498.hg.1 | ENST00000322536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100166498.hg.1 | ENST00000526794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100166498.hg.1 | NM_004398.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100166498.hg.1 | TC1100008991.hg.1 | Clariom D TC |

DDX10
Genomic Map