| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012109.hg.1 |
TC08000938.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012109.hg.1 |
ENST00000330777.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012109.hg.1 |
ENST00000520004.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012109.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012108.hg.1 |
TC08000938.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012108.hg.1 |
ENST00000330777.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012108.hg.1 |
ENST00000520004.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08012108.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009511.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800164970.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164970.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800164970.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164970.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800164970.hg.1 |
TC0800009511.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800164969.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164969.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164969.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800164969.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164969.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164969.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800164969.hg.1 |
TC0800009511.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800164968.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164968.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164968.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800164968.hg.1 |
ENST00000615670.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164968.hg.1 |
ENST00000622241.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800164968.hg.1 |
NM_001080826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800164968.hg.1 |
TC0800009511.hg.1 |
Clariom D TC |