| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000373484.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000409190.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000417633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000417633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000417633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000426909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
ENST00000426909.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_001161708.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013623.hg.1 |
NM_030786.2 |
Refseq |