Assay Details
Gene Symbol: B4GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159387.1 | NP_001152859.1 | 2 - 3 | 240 | 75 | |
NM_001159388.1 | NP_001152860.1 | 2 - 3 | 253 | 75 | ||
NM_153446.2 | NP_703147.2 | 2 - 3 | 455 | 75 | ||
XM_011524314.2 | XP_011522616.1 | 2 - 3 | 608 | 75 | ||
XM_017024173.1 | XP_016879662.1 | 2 - 3 | 302 | 75 | ||
GenBank mRNA | AF510036.1 | - | 2 - 3 | 455 | 75 | |
AJ517770.1 | - | 2 - 3 | 455 | 75 | ||
AJ517771.1 | - | 2 - 3 | 237 | 75 | ||
AK302876.1 | - | 2 - 3 | 253 | 75 | ||
BC113675.1 | - | 2 - 3 | 449 | 8322727 | 75 | |
BC113677.1 | - | 2 - 3 | 449 | 8322729 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000638.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17007971.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007971.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007971.hg.1 | TC17000638.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17007968.hg.1 | ENST00000504681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007968.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007968.hg.1 | TC17000638.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17007973.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007973.hg.1 | ENST00000393354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007973.hg.1 | ENST00000504681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17007973.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007973.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007973.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17007973.hg.1 | TC17000638.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160586.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160586.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160586.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160586.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160586.hg.1 | TC1700008191.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700160585.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160585.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160585.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160585.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160585.hg.1 | TC1700008191.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700160581.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160581.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160581.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160581.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160581.hg.1 | TC1700008191.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700160589.hg.1 | TC1700008191.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 139877 | - | - |
